missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gellan Gum
An agar substitute and gelling agent | CAS: 71010-52-1 | (C24H38O20)A
$225.96 - $658.47
Chemical Identifiers
| CAS | 71010-52-1 |
|---|---|
| Molecular Formula | (C24H38O20)A |
| MDL Number | MFCD00131909 |
| Synonym | Phytagel |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAJ6342330
|
Thermo Scientific Chemicals
J6342330 |
250 g |
Each for $225.96
|
|
|||||
|
AAJ63423A1
|
Thermo Scientific Chemicals
J63423A1 |
1 kg |
Each for $658.47
|
|
|||||
Description
Gellan Gum is an agar substitute and gelling agent, and useful in a wide variety of immobilization matricies.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsGellan Gum is an agar substitute and gelling agent. It is used in the food industry as an anti-settling agent, thickener or stabilizer. Also used in cosmetic industries due to its special gelling property. It is also used in pharmaceuticals and in tissue engineering. It is used as a substitute for agar in leptospiral media. Also useful in a wide variety of immobilization matricies.
Solubility
Soluble in water.
Notes
Incompatible with oxidizing agents.
Chemical Identifiers
| 71010-52-1 | |
| (C24H38O20)A | |
| MFCD00131909 | |
| Phytagel |
Specifications
| 71010-52-1 | |
| (C24H38O20)A | |
| 250 g | |
| Phytagel | |
| Powder |
| White | |
| MFCD00131909 | |
| 14,4383 | |
| Soluble in water. | |
| Gellan Gum |
Safety and Handling
EINECSNumber : 275-117-5
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only