missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
MilliporeSigma™ GelRed™ Nucleic Acid Stain 10000X Water
GelRed™ is a fluorescent nucleic acid stain designed to replace the highly toxic ethidium bromide (EtBr) for staining dsDNA, ssDNA or RNA in agarose gels or polyacrylamide gels.
Supplier: MilliporeSigma™ SCT123
Description
- GelRed™ is a sensitive, stable and environmentally safe fluorescent nucleic acid dye designed to replace the highly toxic ethidium bromide (EtBr) for staining dsDNA, ssDNA or RNA in agarose gels or polyacrylamide gels
- GelRed™ and EtBr have virtually the same spectra, so you can directly replace EtBr with GelRed™ without changing your existing imaging system
- More sensitive than EtBr
- The dye is noncytotoxic and non-mutagenic at concentrations well above the working concentrations used in gel staining
- GelRed™ successfully passed environmental safety tests in compliance with CCR Title 22 Hazardous Waste Characterization, under which GelRed™ is not classified as hazardous waste
- Absorbance: Standard Transilluminator (302 or 312 nm)
- Emission: Ethidium Bromide, SYBR or GelStar Filter
Specifications
Absorbance: Standard Transilluminator (302 or 312 nm); Emission: Ethidium Bromide, SYBR or GelStar Filter | |
Liquid | |
Store at room temperature, protected from light |
Fluorescence | |
0.5 mL |