missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gemcitabine, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 456890050

 View more versions of this product

Catalog No. AC456890050


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Gemcitabine
292.0°C
Authentic
C9H11F2N3O4
gemcitabine, 2',2'-difluorodeoxycytidine, gemcitabinum, gamcitabine, gemcitabina, gemcitabine hcl, dfdc, 2'-deoxy-2',2'-difluorocytidine, folfugem, gemcel
C1=CN(C(=O)N=C1N)C2C(C(C(O2)CO)O)(F)F
263.2
CHEBI:175901
98%
95058-81-4
2% max.
97.5% min. (HPLC)
5 g
SDUQYLNIPVEERB-QPPQHZFASA-N
4-amino-1-[(2R,4R,5R)-3,3-difluoro-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidin-2-one
60750
263.2

Chemical Identifiers

95058-81-4
263.2
gemcitabine, 2',2'-difluorodeoxycytidine, gemcitabinum, gamcitabine, gemcitabina, gemcitabine hcl, dfdc, 2'-deoxy-2',2'-difluorocytidine, folfugem, gemcel
CHEBI:175901
C1=CN(C(=O)N=C1N)C2C(C(C(O2)CO)O)(F)F
C9H11F2N3O4
SDUQYLNIPVEERB-QPPQHZFASA-N
60750
4-amino-1-[(2R,4R,5R)-3,3-difluoro-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidin-2-one
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Suspected of damaging fertility.
Suspected of damaging the unborn child.
May cause damage to organs through prolonged or repeated exposure if inhaled.<br

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Take off contaminated clothing and wash before reuse.
IF IN EYES: Rinse cautiously with water

GHS Signal Word: Warning

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only