Epredia™ Gemini™ ClearVue™ Combined Staining and Coverslipping Solution
Paired together by the Epredia™ Slide Basket Transfer System, the Gemini™ AS Slide Stainer and ClearVue™ Coverslipper eliminate downtime and increase laboratory productivity.
Specifications
For Use With (Application) | Automatic Transfer from the Gemini AS Slide Stainer to the ClearVue Coverslipper |
---|---|
Height (Metric) | 89 cm |
Electrical Requirements | 100/240 V 50/60 Hz |
Catalog Number | Mfr. No. | Includes | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Includes | Price | Quantity | |||||
A84500001
|
Epredia™
A84500001 |
Slide Basket Transfer System |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
- Free up technical time with the automatic transfer of slide baskets from the Gemini AS Slide Stainer to the ClearVue Coverslipper by the Slide Basket Transfer System
- Simply place slides in the Gemini AS slide stainer and carry on with other tasks free of distractions
- System will sound an alert when the slides have finished coverslipping in the ClearVue Coverslipper
- Reduce exposure to harmful fumes by eliminating the transfer between the Gemini AS Slide Stainer and ClearVue Coverslipper
- Due to the unique layout of the Gemini Slide Stainer, the combination solution has a space-saving design compared to other combination units
- System is available with either a heated Gemini AS Stainer (A84500005) for histology laboratories, or with a non-heated Gemini AS Stainer (A84500006) for cytology laboratories
- Slide Basket Transfer System (A84500001) can be purchased separately and is compatible for existing customers with Gemini AS and ClearVue instruments manufactured after February 2018
Specifications
Automatic Transfer from the Gemini AS Slide Stainer to the ClearVue Coverslipper | |
89 cm | |
100/240 V 50/60 Hz |