missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ GeneArt™ Genomic Cleavage Detection Kit DFS Item

The GeneArt Genomic Cleavage Detection Kit provides a simple, reliable, and rapid method for the detection of locus-specific double-strand break formation.

Supplier:  Invitrogen™ A24372

Catalog No. A24372


Explore available promotions

Includes:
  • 1 bottle Cell Lysis Buffer
  • 1 tube Proteinase K
  • 1 tube PCR Supermix
  • 1 tube Water
  • 1 tube Detection Enzyme
  • 1 tube Detection Reaction Buffer
  • 1 tube Control Template & Primers

Description

Description

The GeneArt Genomic Cleavage Detection Kit provides a simple, reliable, and rapid method for the detection of locus-specific double-strand break formation. When using genome editing tools to obtain targeted mutations, it is necessary to determine the efficiency at which these nucleases cleave the target sequence, particularly prior to proceeding to the more laborious and expensive processes of cloning and sequencing.

The GeneArt genomic cleavage detection kit contains all of the reagents required prior to gel analysis. A sample of the edited cell population is used as a direct PCR template for amplification with primers specific to the targeted region. The PCR product is then denatured and re-annealed to produce heteroduplex mismatches where double strand breaks have occurred, resulting in indel introduction. These mismatches are recognized and cleaved by the detection enzyme. This cleavage is both easily detectable and quantifiable using gel analysis.

  • Easy—no genomic DNA isolation, direct PCR amplification
  • Rapid—4 hour total processing time
  • Quantative—gel band density directly correlated to target indel introduction

Recommended Storage

Store all at -5 to -30°C

Specifications

Specifications

Contains:

• 1 bottle Cell Lysis Buffer
• 1 tube Proteinase K
• 1 tube PCR Supermix
• 1 tube Water
• 1 tube T7E1 Detection Enzyme
• 1 tube T7E1 Detection Reaction Buffer
• 1 tube Control Template & Primers

Store all components at -5° to -30°C.
Liquid
Taq Polymerase
CRISPR-Cas9, TAL Effector Nuclease
20 reactions
20 Reactions
Primer-probe
Kit
Fast
Genomic Cleavage Detection Kit
INDEL
20 Reactions
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.