Genecon™ Hand-Held Generator
Durable for long-time classroom use.
Supplier:  Customer Channels Group (Ccg) B2631
Catalog No. S43917
Explore available promotions
Description
Description
One attachable parallel electric cord with two covered alligator clips is included.
  • Made from transparent ABS resin (blue)
  • Dimensions: 5.6 × 4.6 × 1.6 in. (14 × 11.5 × 4cm)
Specifications
Specifications
High school students can learn to analyze the relationship between an electric current and the strength of its magnetic field using simple electromagnets, college students can review sophisticated principles, such as Ohm's Law and the electromagnetic properties of light
One attachable parallel electric cord with two covered alligator clip, A/B identification stickers
0.115 kg
12 VDC
0.253 lb.