General Biology Slide Set A
Prepared slides of animal and plant structures in whole mount (w.m.), longitudinal section (l.s.) and cross-section (c.s.) preparations.
Supplier:  Customer Channels Group (Ccg) EPDS500
Catalog No. S500
Explore available promotions
Description
Description
Set A offers prepared slides of animal and plant structures in whole mount (w.m.), longitudinal section (l.s.) and cross-section (c.s.) preparations. Also available in CD-ROM format (see S500-CD, at left).
Zoology
  • Amoeba proteus, w.m., showing nucleus and pseudopodia
  • Hydra, w.m., extended, to show foot, body, mouth, and tentacles
  • Lumbricus, c.s., back of clitellum showing muscular wall, intestine, typhlosole, and nephridia
  • Daphnia and Cyclops, w.m., both specimens on slide
  • Musca domestica, leg, fly with clinging pads (pulvilli)
  • Apis mellifica, honeybee anterior and posterior wing
  • Squamous epithelium, isolated (simple) from human cheek
  • Striated muscle, l.s., showing nuclei and striations
  • Compact bone, c.s., specially stained for lamellae, canaliculi
  • Human scalp, l.s., showing hair follicles, sebaceous glands, epidermis
  • Human blood smear, stained for red and white corpuscles
Bacteria and Cryptogams
  • Bacteria from mouth, smear, Gram stained showing bacilli, cocci, spirilli, and spirochaetes
  • Diatoms, strewn slide of mixed species
  • Spirogyra, vegetative filaments with spiral chloroplasts. Rhizopus, w.m., (or Mucor), mycelium and sporangia
  • Moss, w.m., stem with leaves
Botany: Phanerogams Slides
  • Rannunculus, c.s., buttercup, typical dicot root
  • Zea mays, c.s., corn, monocot stem with bundles
  • Helianthus, c.s., sunflower, herbaceous dicot stem
  • Syringa, c.s., lilac leaf, showing epidermis, palisade parenchyma, spongy parenchyma, and vascular bundles
  • Lilium, c.s., lily, anthers with pollen grains and pollen sacs
  • Lilium, c.s., ovary showing arrangement of ovules
  • Allium, w.m., onion epidermis with cell nuclei
  • Allium, l.s., onion root tip for mitosis, cell division
Specifications
Specifications
Animal and Plant Structures
25/Pk.