Surgical Design™ General Purpose Lab Spatula
Fisherbrand™ General Purpose Lab Spatula is used to transfer sticky or corrosive materials without scratching or damaging container surfaces
 View more versions of this product
Catalog No. 13-812-207
Explore available promotions
Description
Description
  • Non-stick coating
  • Multiple sizes allow for varied laboratory applications.
Specifications
Specifications
1
General Purpose Spatula
6 in.
Stainless Steel
PTFE
Non-stick lab applications
Wooden Handle
Product Suggestions
Product Suggestions