Thermo Scientific™ GeneRuler DNA Ladder Mix, ready-to-use
Perform sizing and approximate quantification of a wide range of double-stranded DNA on agarose gels.
Supplier: Thermo Scientific™ SM0334
Description
GeneRuler DNA Ladder Mix, ready-to-use, is recommended for sizing and approximate quantification of a wide range of double-stranded DNA on agarose gels. The ladder is a mixture of chromatography-purified individual DNA fragments.
GeneRuler DNA Ladder Mix, ready-to-use, is premixed with 6X DNA Loading Dye for direct loading on gel. The ladder is supplied with 6X DNA Loading Dye.
Highlights
- Ideal for both DNA sizing and approximate quantification.
- Sharp bands.
- Bright reference bands (given in red).
- Can be directly loaded and are stable at room temperature for 6Â months.
- Supplied with loading dye for sample DNA.
Specifications
6X | |
Xylene Cyanol FF, Bromophenol Blue, Orange G | |
Yes | |
Agarose Gels | |
GeneRuler |
Agarose | |
DNA Ladder Mix | |
100 to 1000 bp | |
Sustainable packaging | |
50 μg, 100 applications |
For Research Use Only. Not for use in diagnostic procedures.