Thermo Scientific™ GENESYS™ 30 Visible Spectrophotometer Accessories
Get all of the tools you need to facilitate your analytical process with accessories for the Thermo Scientific™ GENESYS™ 30 Visible Spectrophotometer.
Supplier:  Thermo Scientific™ 840259800
 View more versions of this product
Catalog No. 14-380-005
Explore available promotions
Description
Description
Get all of the tools you need to facilitate your analytical process with accessories for the Thermo Scientific™ GENESYS™ 30 Visible Spectrophotometer.
  • Package of 10 rolls
  • 80mm width
  • Use with thermal printers for:
    •   GENESYS 30
    •   GENESYS 10S
    •   BioMate 3S
    •   Evolution 60S
    •   SPECTRONIC 200/200E
Specifications
Specifications
Self Stick Printer Paper
Print your samples and adhere them to your lab booklet without the need to staples or tape.
10 rolls of paper
Self-Stick Printer Paper
Thermal printers for: GENESYS 30 and 10S, BioMate 3S, Evolution 60S, SPECTRONIC 200/200E