Thermo Scientific™ GENESYS™ 30 Visible Spectrophotometer Accessories
Get all of the tools you need to facilitate your analytical process with accessories for the Thermo Scientific™ GENESYS™ 30 Visible Spectrophotometer.
Supplier:  Thermo Scientific™ 840278000
 View more versions of this product
Catalog No. 14-380-453
Explore available promotions
Description
Description
Get all of the tools you need to facilitate your analytical process with accessories for the Thermo Scientific™ GENESYS™ 30 Visible Spectrophotometer.
  • Printer attaches directly to instrument behind screen
  • Easy drop-in paper re-loading
  • Uses 80mm wide thermal paper rolls
Specifications
Specifications
This snap-on thermal printer attaches easily anddirectly to the back of the instrument and uses 80mm wide thermal drop in paper rolls. It can also be used with regular or self stick printer paper.
Direct hard-copy printing of results from the instrument onboard software control.
1 roll thermal paper
Thermal Snap-on Printer
GENESYS 30 Visible Spectrophotometer
SDS
Documents
Documents
Accessories