missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Genistin, 99+%, Thermo Scientific Chemicals

Glucoside of genistein that inhibits protein tyrosine kinase

$865.68 - $865.68

Chemical Identifiers

CAS 529-59-9
Molecular Formula C21H20O10
Molecular Weight (g/mol) 432.381
MDL Number MFCD00016883
InChI Key ZCOLJUOHXJRHDI-CMWLGVBASA-N
Synonym genistin, genistine, genisteol 7-monoglucoside, genistoside, genistein 7-glucoside, glucosyl-7-genistein, genistein 7-o-beta-d-glucoside, unii-1pog3scn5t, genistein glucoside, genistein-7-glucoside
PubChem CID 5281377
ChEBI CHEBI:27514
IUPAC Name 5-hydroxy-3-(4-hydroxyphenyl)-7-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxychromen-4-one
SMILES C1=CC(=CC=C1C2=COC3=CC(=CC(=C3C2=O)O)OC4C(C(C(C(O4)CO)O)O)O)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ63445MC
View Documents
Thermo Scientific Chemicals
J63445MC
100 mg
Each for $865.68
 
Description

Description

Glucoside of genistein that inhibits protein tyrosine kinase

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

529-59-9
432.381
ZCOLJUOHXJRHDI-CMWLGVBASA-N
5281377
5-hydroxy-3-(4-hydroxyphenyl)-7-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxychromen-4-one
C21H20O10
MFCD00016883
genistin, genistine, genisteol 7-monoglucoside, genistoside, genistein 7-glucoside, glucosyl-7-genistein, genistein 7-o-beta-d-glucoside, unii-1pog3scn5t, genistein glucoside, genistein-7-glucoside
CHEBI:27514
C1=CC(=CC=C1C2=COC3=CC(=CC(=C3C2=O)O)OC4C(C(C(C(O4)CO)O)O)O)O

Specifications

529-59-9
Brown to Yellow
MFCD00016883
64479
genistin, genistine, genisteol 7-monoglucoside, genistoside, genistein 7-glucoside, glucosyl-7-genistein, genistein 7-o-beta-d-glucoside, unii-1pog3scn5t, genistein glucoside, genistein-7-glucoside
C1=CC(=CC=C1C2=COC3=CC(=CC(=C3C2=O)O)OC4C(C(C(C(O4)CO)O)O)O)O
432.381
CHEBI:27514
≥99%
Genistin
258°C to 259°C
C21H20O10
100 mg
14,4391
ZCOLJUOHXJRHDI-CMWLGVBASA-N
5-hydroxy-3-(4-hydroxyphenyl)-7-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxychromen-4-one
5281377
432.38
Powder
Videos
Safety and Handling

Safety and Handling

RTECSNumber : DJ3093000

TSCA : No

Recommended Storage : Store at -20°C

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only