Genlantis™ NeuroPure™ E18 Primary Rat Cortical Cells DFS Item
Save time with fresh, ready-to-use cells
Supplier:  Genlantis™ N200200
Catalog No. N200200
Explore available promotions
Includes: Day 18 Embryonic Sprague/Dawley Rat Cortical Cells (1 vial x 2mL); NeuroPure™ Plating Medium + 25μM glutamate (1 vial x 12mL); NeuroPrep Medium (1 vial x 2.5mL); NeuroPapain™ Enzyme (1 vial x 5mg)
Description
Description
NeuroPure™ E18 Primary Rat Cortical Cells are live neurons provided as micro-surgically dissected regions of day 18 embryonic Sprague/Dawley rat brain. These cells are prepared fresh each week and shipped in a nutrient rich medium that keeps them alive for up to 14 days under refrigeration. Following simple dissociation steps, the NeuroPure cells can be quickly plated on almost any poly-lysine coated substrate using the provided NeuroPure Plating Medium.
  • Freshly isolated healthy cells—never frozen
  • Get your primary culture up and running within one hour
  • Pure neuronal cells, 99.9% glial cell free
  • Isolated from microsurgically dissected regions of Sprague/Dawley or Fischer 344 rats
  • Available in seven popularly used cell types
Transfection, Pharmacology, Electrophysiology, Immunocytochemistry, and Neuronal Development Studies.
Specifications
Specifications
4° to 8°C
Rat
Day 18 embryonic sprague/dawley Rat cortical cells (1 vial x 2mL), NeuroPure™ plating medium + 25μM glutamate (1 vial x 12mL), NeuroPrep™ medium (1 vial x 2.5mL), NeuroPapain™ Enzyme
Rat Cortical Cells
Transfection, pharmacology, electrophysiology, immunocytochemistry and neuronal development studies
2 Million Cells
Product Suggestions
Product Suggestions