Genlantis™ PrimaPure™ Preadipocytes Kits
High-quality human preadipocytes from adult heart or subcutaneous tissue
Supplier: Genlantis™ PH80205AK
Specifications
4° and -20°C | |
Study insulin-stimulated glucose transport, growth hormone enhanced lypolysis and obese gene expression | |
Human Preadipocyte Growth Medium | |
Human Adipose Tissue |
Human Preadipocytes (Heart, Adult) | |
One ampoule of cryopreserved HPAd, 500mL of Human preadipocyte growth medium, one subculture reagent kit (100mL each of HEPES buffered saline solution, trypsin/EDTA, and trypsin neutralizing solution) | |
1 Kit | |
16 Population Doublings |