Genlantis™ SoluBL21 Competent E. coli
Supplier: Genlantis™ C700200
Description
SoluBL21™ Competent E. coli, a significantly improved BL21 host strain for toxic and soluble protein expression. Click on the DATA tab. Using a directed evolution approach, Genlantis scientists have developed a mutant strain of BL21(DE3) E. coli that can produce soluble protein in the majority of cases where expression in the parent BL21(DE3) yielded no detectable soluble product.
- Express toxic proteins
- Optimized E. coli strain for expressing insoluble proteins in soluble form
- Simple and straightforward protocol
- Compatible with all T7 promoter-based expression vectors
- 2 White Papers
Specifications
SoluBL21 Competent E. coli | |
SoluBL21 chemically competent E. coli 10 x 50μL, SOC medium 5mL, pUC19 positive control Plasmid 20μl (500pg/μl) | |
Each |