Amsbio LLC (formerly Genlantis) Zeus™ Electroporation Cuvets
Compatible with all common electroporation devices
Supplier: Amsbio LLC (formerly Genlantis) C904050
Description
- Individually wrapped and gamma-irradiated to ensure sterility
- Gap tolerance of 0.02 mm to ensure reproducible results
- Color-coded, leak-resistance caps for easy identification of gap width
- Provide consistent electroporation results
- Compatible with all common electroporation devices, and are offered at an economical price
Specifications
Sterile | |
Electroporation Cuvet | |
4mm | |
Electorporation of Bacteria, Yeast, Mammalian Cells |