Learn More
Geraniol, 97%, Thermo Scientific Chemicals
$64.62 - $821.25
Chemical Identifiers
CAS | 106-24-1 |
---|---|
Molecular Formula | C10H18O |
Molecular Weight (g/mol) | 154.253 |
MDL Number | MFCD00002917 |
InChI Key | GLZPCOQZEFWAFX-JXMROGBWSA-N |
Synonym | geraniol, lemonol, geranyl alcohol, trans-geraniol, e-geraniol, e-nerol, trans-3,7-dimethyl-2,6-octadien-1-ol, e-3,7-dimethylocta-2,6-dien-1-ol, 2e-3,7-dimethylocta-2,6-dien-1-ol, geraniol extra |
PubChem CID | 637566 |
ChEBI | CHEBI:17447 |
IUPAC Name | (2E)-3,7-dimethylocta-2,6-dien-1-ol |
SMILES | CC(=CCCC(=CCO)C)C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1373618
|
Thermo Scientific Chemicals
A1373618 |
50 g |
Each for $64.62
|
|
|||||
AAA1373630
|
Thermo Scientific Chemicals
A1373630 |
250 g |
Each for $263.01
|
|
|||||
AAA137360B
|
Thermo Scientific Chemicals
A137360B |
1000 g |
Each for $821.25
|
|
|||||
Description
Geraniol is used in the synthesis of insect repellant. It is also used in the synthesis of Angelicoin A and Herecinone J, which inhibit collagen-induced platelet aggregation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsGeraniol is used in the synthesis of insect repellant. It is also used in the synthesis of Angelicoin A and Herecinone J, which inhibit collagen-induced platelet aggregation.
Solubility
Not miscible in water.
Notes
Incompatible with Strong oxidizing agents, acid chlorides, acid anhydrides. Keep away from heat. Air sensitive.
Chemical Identifiers
106-24-1 | |
154.253 | |
GLZPCOQZEFWAFX-JXMROGBWSA-N | |
637566 | |
(2E)-3,7-dimethylocta-2,6-dien-1-ol |
C10H18O | |
MFCD00002917 | |
geraniol, lemonol, geranyl alcohol, trans-geraniol, e-geraniol, e-nerol, trans-3,7-dimethyl-2,6-octadien-1-ol, e-3,7-dimethylocta-2,6-dien-1-ol, 2e-3,7-dimethylocta-2,6-dien-1-ol, geraniol extra | |
CHEBI:17447 | |
CC(=CCCC(=CCO)C)C |
Specifications
106-24-1 | |
0.884 | |
101°C (213°F) | |
C10H18O | |
MFCD00002917 | |
1722456 | |
geraniol, lemonol, geranyl alcohol, trans-geraniol, e-geraniol, e-nerol, trans-3,7-dimethyl-2,6-octadien-1-ol, e-3,7-dimethylocta-2,6-dien-1-ol, 2e-3,7-dimethylocta-2,6-dien-1-ol, geraniol extra | |
GLZPCOQZEFWAFX-JXMROGBWSA-N | |
(2E)-3,7-dimethylocta-2,6-dien-1-ol | |
637566 | |
154.25 | |
Geraniol |
-15°C | |
114°C to 115°C (12 mmHg) | |
Floral | |
1.476 | |
50 g | |
14,4403 | |
Not miscible in water. | |
CC(=CCCC(=CCO)C)C | |
154.253 | |
CHEBI:17447 | |
97% |
Safety and Handling
GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
P210-P261-P264b-P272-P280-P302+P352-P305+P351+P338-P310-P333+P313-P362-P370+P378q-P501c
H227-H315-H317-H318
EINECSNumber : 203-377-1
RTECSNumber : RG5830000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only