missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Germanium dioxide, 99.999%, (trace metal basis), electronic grade, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 190002500

 View more versions of this product

Catalog No. AC190002500


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis
Specifications

Specifications

Germanium Dioxide
1310-53-8
Powder
99.999%
GeO2
15,4443
Solubility in water: 0.4%. Other solubilities: soluble in about 100 parts boiling water, soluble in acids or solutions of the fixed, alkalies
O=[Ge]=O
104.59
104.59
Trace Metal Basis
1115.0°C
White
250 g
GeO2
MFCD00011030
germanium dioxide, germania, germanic acid, germanium oxide, germanium iv oxide, germanic oxide crystalline, dioxogermanium, germanic oxide, wln: ge o2
YBMRDBCBODYGJE-UHFFFAOYSA-N
dioxogermane
14796
99.999%
Glass bottle

Chemical Identifiers

1310-53-8
104.59
YBMRDBCBODYGJE-UHFFFAOYSA-N
14796
O=[Ge]=O
GeO2
MFCD00011030
germanium dioxide, germania, germanic acid, germanium oxide, germanium iv oxide, germanic oxide crystalline, dioxogermanium, germanic oxide, wln: ge o2
dioxogermane
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Harmful if inhaled.

GHS P Statement
Wear protective gloves/eye protection/face protection.
Wash face,hands and any exposed skin thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWAL

GHS Signal Word: Warning

EINECSNumber : 215-180-8

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only