Learn More
Germanium(IV) chloride, 99.9999% (metals basis), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 01050914
Description
It is used as a catalyst in the conversion of carbohydrates into 5-hydroxymethylfurfural in ionic liquids, as a reducing agent in combination with triphenylphosphine (TPP) for the reduction of alpha-bromo carboxylic acid derivatives, and in the production of pure germanium. It also finds use as an intermediate for several optical processes. It is one of the most important dopants in silica glass for optical fibers. It is employed in the preparation of germanium dioxide which is used for wide camera lens, microscopy, IR-transparent glasses/windows/lenses, and for the core of fiber-optic lines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
83.1°C | |
-49.5°C | |
Colorless | |
25 g | |
Cl4GeH8 | |
MFCD00011029 | |
14,4410 | |
Soluble in ether,benzene,chloroform,carbon tetrachloride. Insoluble in hydrochloric acid and sulphuric acid. | |
Cl.Cl.Cl.Cl.[GeH4] | |
222.49 | |
214.4 | |
Acidic | |
1.874 g/mL |
Germanium(IV) chloride | |
10038-98-9 | |
Liquid | |
(metals basis) | |
GeCl4 | |
UN3264 | |
germanium tetrachloride, germanium chloride, germane, tetrachloro, germanium iv chloride, germanium, tetrachloro, gecl4, germanium chloride gecl4, unii-ysv1r803c0, wln: .ge..g4, germanium chloride gecl4 8ci | |
VDHBHZZBEGQJRK-UHFFFAOYSA-N | |
tetrachlorogermane | |
66226 | |
99.9999% | |
Moisture sensitive |
Chemical Identifiers
10038-98-9 | |
222.49 | |
VDHBHZZBEGQJRK-UHFFFAOYSA-N | |
66226 |
Cl4GeH8 | |
MFCD00011029 | |
germanium tetrachloride, germanium chloride, germane, tetrachloro, germanium iv chloride, germanium, tetrachloro, gecl4, germanium chloride gecl4, unii-ysv1r803c0, wln: .ge..g4, germanium chloride gecl4 8ci | |
Cl.Cl.Cl.Cl.[GeH4] |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c
H290-H314-H335
DOTInformation : Hazard Class: 8; Packaging Group: I
EINECSNumber : 233-116-7
RTECSNumber : LY5225000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only