missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Germanium pieces, 3-9mm (0.1-0.4in), 99.999% (metals basis), Thermo Scientific Chemicals

$385.77 - $1365.05

Chemical Identifiers

CAS 7440-56-4
Molecular Formula Ge
Molecular Weight (g/mol) 72.63
MDL Number MFCD00085310
InChI Key GNPVGFCGXDBREM-UHFFFAOYSA-N
Synonym element, germanio, germanium, metal powder, unii-00072j7xws, powder, germanium, chips trace metals basis, atom, copper metal, germanium, disks, 15mm, thickness 1.0mm, polycrystalline, germanium, powder
PubChem CID 6326954
ChEBI CHEBI:30441
IUPAC Name germanium
SMILES [Ge]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1019109
View Documents
Thermo Scientific Chemicals
01019109
10 g
Each for $385.77
 
AA1019118
View Documents
Thermo Scientific Chemicals
01019118
50 g
Each for $1,365.05
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7440-56-4
72.63
GNPVGFCGXDBREM-UHFFFAOYSA-N
6326954
germanium
Ge
MFCD00085310
element, germanio, germanium, metal powder, unii-00072j7xws, powder, germanium, chips trace metals basis, atom, copper metal, germanium, disks, 15mm, thickness 1.0mm, polycrystalline, germanium, powder
CHEBI:30441
[Ge]

Specifications

7440-56-4
MFCD00085310
element, germanio, germanium, metal powder, unii-00072j7xws, powder, germanium, chips trace metals basis, atom, copper metal, germanium, disks, 15mm, thickness 1.0mm, polycrystalline, germanium, powder
[Ge]
72.63
CHEBI:30441
Odorless
Germanium pieces, 3 to 9mm (0.1 to 0.4 in.)
Ge
10 g
GNPVGFCGXDBREM-UHFFFAOYSA-N
germanium
6326954
99.999%
Pieces
(metals basis)
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 231-164-3

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only