missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Gestodene, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 468652500
Specifications
Gestodene | |
Glass Bottle | |
White to Beige | |
C21H26O2 | |
4425 | |
SIGSPDASOTUPFS-KQMXEUTGSA-N | |
(1R)-11a-ethyl-1-ethynyl-1-hydroxy-1H,3aH,3bH,4H,5H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-7-one | |
310.43 | |
Powder |
60282-87-3 | |
250 mg | |
Available | |
00867858 | |
13beta-Ethyl-17beta-hydroxy-18,19-dinorpregna-4,15-dien-20-yn-3-one | |
CCC12CCC3C(CCC4=CC(=O)CCC34)C1C=C[C@@]2(O)C#C | |
310.44 | |
197.9°C |
Chemical Identifiers
60282-87-3 | |
310.44 | |
SIGSPDASOTUPFS-KQMXEUTGSA-N | |
(1R)-11a-ethyl-1-ethynyl-1-hydroxy-1H,3aH,3bH,4H,5H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-7-one |
C21H26O2 | |
00867858 | |
13beta-Ethyl-17beta-hydroxy-18,19-dinorpregna-4,15-dien-20-yn-3-one | |
CCC12CCC3C(CCC4=CC(=O)CCC34)C1C=C[C@@]2(O)C#C |
Safety and Handling
GHS08: Health hazard
ShelfLife : 5 years
EINECSNumber : 262-145-8
RTECSNumber : JF7956000
Recommended Storage : Freezer -20°C