Learn More
Gibberellic acid, 99%
CAS: 77-06-5 | C19H22O6 | 346.38 g/mol
$126.46 - $1,343.05
Chemical Identifiers
| CAS | 77-06-5 |
|---|---|
| Molecular Formula | C19H22O6 |
| Molecular Weight (g/mol) | 346.38 |
| MDL Number | MFCD00079329 |
| InChI Key | IXORZMNAPKEEDV-QTWFBFKQSA-N |
| Synonym | gibberellic acid, gibberellin a3, gibberellin, gibreskol, brellin, cekugib, grocel, gibberellin x, gibberellic acid ga3, gib-tabs |
| PubChem CID | 91757643 |
| IUPAC Name | (1R,2R,5S,8S,9S,10R,12S)-5,12-dihydroxy-11-methyl-6-methylidene-16-oxo-15-oxapentacyclo[9.3.2.1âµ,â¸.0¹,¹â°.0²,â¸]heptadec-13-ene-9-carboxylic acid |
| SMILES | CC12[C@H]3[C@H](C(O)=O)[C@@]45CC(=C)[C@@](O)(C4)CC[C@H]5[C@]3(OC1=O)C=C[C@@H]2O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC410910010
|
Thermo Scientific Chemicals
410910010 |
1 g | Glass bottle |
N/A
|
|
||||
|
AC410910050
|
Thermo Scientific Chemicals
410910050 |
5 g | Glass bottle |
N/A
|
|
||||
|
AC410910250
|
Thermo Scientific Chemicals
410910250 |
25 g | Glass bottle |
N/A
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 77-06-5 | |
| 346.38 | |
| IXORZMNAPKEEDV-QTWFBFKQSA-N | |
| 91757643 | |
| CC12[C@H]3[C@H](C(O)=O)[C@@]45CC(=C)[C@@](O)(C4)CC[C@H]5[C@]3(OC1=O)C=C[C@@H]2O |
| C19H22O6 | |
| MFCD00079329 | |
| gibberellic acid, gibberellin a3, gibberellin, gibreskol, brellin, cekugib, grocel, gibberellin x, gibberellic acid ga3, gib-tabs | |
| (1R,2R,5S,8S,9S,10R,12S)-5,12-dihydroxy-11-methyl-6-methylidene-16-oxo-15-oxapentacyclo[9.3.2.1âµ,â¸.0¹,¹â°.0²,â¸]heptadec-13-ene-9-carboxylic acid |
Specifications
| 77-06-5 | |
| White to Yellow | |
| 99% | |
| C19H22O6 | |
| 1 g | |
| 82.50 (20.00°C c=10,ethanol) | |
| gibberellic acid, gibberellin a3, gibberellin, gibreskol, brellin, cekugib, grocel, gibberellin x, gibberellic acid ga3, gib-tabs | |
| IXORZMNAPKEEDV-QTWFBFKQSA-N | |
| (1R,2R,5S,8S,9S,10R,12S)-5,12-dihydroxy-11-methyl-6-methylidene-16-oxo-15-oxapentacyclo[9.3.2.1âµ,â¸.0¹,¹â°.0²,â¸]heptadec-13-ene-9-carboxylic acid | |
| 91757643 | |
| 99% | |
| Gibberellic acid |
| 227°C | |
| Authentic | |
| Glass bottle | |
| MFCD00079329 | |
| 15, 4454 | |
| 82.5 | |
| Solubility in water: 5g/L (20° c). Other solubilities: soluble in aq.sol. of sodium bicarbonate,soluble in methanol,ethanol,acetone and aq. sol.,of sodium acetate,moderately soluble in ethyl acetate- slightly,soluble in ether- insoluble in chl | |
| CC12[C@H]3[C@H](C(O)=O)[C@@]45CC(=C)[C@@](O)(C4)CC[C@H]5[C@]3(OC1=O)C=C[C@@H]2O | |
| 346.38 | |
| 346.38 | |
| Crystalline Powder |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Warning
EINECSNumber : 201-001-
RUO – Research Use Only