missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gibco™ Advanced MEM DFS Item

Advanced MEM (Minimum Essential medium) is a widely used basal medium that allows the culture of mammalian cells with reduced Fetal Bovine Serum (FBS) supplementation.

Supplier:  Gibco™ 12492021

 View more versions of this product

Catalog No. 12492021


Explore available promotions

Description

Description

Advanced MEM (Minimum Essential medium) is a widely used basal medium that allows the culture of mammalian cells with reduced Fetal Bovine Serum (FBS) supplementation. Compared to classic MEM, serum supplementation can be reduced by 50-90% with no change in growth rate or morphology.i Cells successfully cultured in Advanced DMEM / F-12, with no adaptation, include PK-15, MDCK, Vero, WI-38 and Hep-2.

This Advanced MEM is manufactured as follows:

With: Glucose, Non-essential Amino Acids, Sodium Pyruvate, Phenol Red

Without: L-glutamine, HEPES

The complete formulation is available.

Gibco™ Advanced MEM is unique from other media due to addition of the following ingredients to allow for serum reduction: ethanolamine, glutathione, ascorbic acid, insulin, transferrin, AlbuMAX™ I lipid-rich bovine serum albumin for cell culture, and the trace elements sodium selenite, ammonium metavanadate, cupric sulfate, and manganous chloride.

Product Use

For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.

Advanced MEM requires supplementation with 1-5% Fetal Bovine Serum and 4mM L-glutamine or GlutaMAX™ supplement. Many cell lines do not require adaptation to this media. The FBS concentration must be optimized for each cell line to obtain maximum serum reduction. Advanced DMEM uses a sodium bicarbonate buffer system (3.7 g/L) and therefore requires a 5-10% CO2 environment to maintain physiological pH.

iPaul Price, Brad Stiles, David Staines, Ethel Evege, Femi Fatunmbi, Kate Wagner, Lori Nestler, David Jayme. Advanced Media as Alternatives to Classical Basal Media. FOCUS, 2003. 25.2:3-6.

Specifications

Specifications

Storage conditions: 2-8° C. Protect from light
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture
Animal Origin
Liquid
Reduced Serum
Sterile-filtered
No Glutamine, No HEPES
Gibco™
Ambient
HeLa, BHK-21, 293, HEP-2, HT-1080, MCF-7, and fibroblasts
1 X
MEM (Minimum Essential Medium)
Sterile-filtered
Low Glucose, Phenol Red, Sodium Pyruvate, Non Essential Amino Acids (NEAA)
cGMP-compliant under the ISO 13485 standard
10 x 500 mL
Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.