Learn More
Gibco™ CMRL Medium, no glutamine
CMRL medium was originally developed by Connaught Medical Research Laboratories for the growth of Earle's 'L'cells under serum-free conditions.
Supplier: Gibco™ 11530037
Requires : Supplementation
Description
CMRL medium was originally developed by Connaught Medical Research Laboratories for the growth of Earle's 'L'cells under serum-free conditions. CMRL medium is also especially useful for cloning monkey kidney cells and for growth of many other mammalian cell lines when supplemented with horse or calf serum.
This CMRL medium is manufactured as follows:
With: Phenol Red
Without: L-glutamine, HEPES
The complete formulation is available.
Gibco™ CMRL medium is a modified version of Medium 199. CMRL medium is particularly rich in nucleosides and vitamins
Product Intended Use
For in vitro diagnostic use. CAUTION: Not for human or animal therapeutic use. Uses other than the labeled intended use may be a violation of local law.
CMRL medium contains no proteins or growth factors. Therefore, CMRL medium requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). CMRL medium uses a sodium bicarbonate buffer system (2.2 g/L) and therefore requires a 5-10% CO2 environment to maintain physiological pH.
Order Info
Shipping Condition: Room Temperature
Specifications
Storage conditions: 2-8°C. Protect from light Shipping conditions: Ambient Shelf life: 12 months from date of manufacture |
|
Monkey Kidney Cells | |
1 X | |
CMRL Medium | |
Low Glucose, Phenol Red | |
cGMP-compliant under the ISO 13485 standard | |
500 mL |
Earle's 'L'cells | |
Animal Origin | |
Liquid | |
Sterile-filtered | |
No Glutamine, No HEPES, No Sodium Pyruvate | |
Gibco™ | |
Room Temperature |
For Research Use Only. Not for use in diagnostic procedures. Not for human or animal therapeutic use. Uses other than the labeled intended use may be a violation of local law. For in vitro diagnostic use.