missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gibco™ DMEM/F-12, HEPES DFS Item

Dulbecco's Modified Eagle Medium:Nutrient Mixture F-12 (DMEM / F-12) is a widely used basal medium for supporting the growth of many different mammalian cells.

$105.00 - $982.00

Specifications

Classification Animal Origin-free
Concentration 1 X
Form Liquid
Product Type DMEM (Dulbecco's Modified Eagle Medium)/F-12
Sterility Sterile-filtered
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
11330032 DFS Item
View Documents
Gibco™
11330032
500 mL
Each for $105.00
 
11330057 DFS Item
View Documents
Gibco™
11330057
10 x 500 mL
Each for $842.00
 
11330099 DFS Item
View Documents
Gibco™
11330099
5 L
Each for $714.00
 
11330107 DFS Item
View Documents
Gibco™
11330107
10 L
Each for $982.00
 
Description

Description

Dulbecco's Modified Eagle Medium:Nutrient Mixture F-12 (DMEM / F-12) is a widely used basal medium for supporting the growth of many different mammalian cells. Cells successfully cultured in DMEM / F-12 include MDCK, glial cells, fibroblasts, human endothelial cells, and rat fibroblasts. We offer a variety of Gibco™ DMEM / F-12 modifications for a range of cell culture applications. Find the right formulation using the media selector tool.

This DMEM / F-12 is modified as follows:

With: L-glutamine, HEPES, Phenol Red

The complete formulation is available.Gibco™ DMEM / F-12 is a 1:1 mixture of DMEM and Ham's F-12. This formulation combines DMEM's high concentrations of glucose, amino acids, and vitamins with F-12's wide variety of components.

DMEM / F-12 contains no proteins, lipids, or growth factors. Therefore, DMEM / F-12 may require supplementation, commonly with 10% Fetal Bovine Serum (FBS). DMEM / F-12 uses a sodium bicarbonate buffer system and therefore requires a 5-10% CO2 environment to maintain physiological pH.

Order Info

Shipping Condition: Room Temperature

Specifications

Specifications

Animal Origin-free
Liquid
Sterile-filtered
High Glucose, Glutamine, HEPES, Phenol Red, Sodium Pyruvate
1 X
DMEM (Dulbecco's Modified Eagle Medium)/F-12
Sterile-filtered
cGMP-compliant under the ISO 13485 standard
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.