Learn More
Gibco™ DMEM/F-12, HEPES, no phenol red
DMEM/F-12 (Dulbecco's Modified Eagle Medium/Nutrient Mixture F-12) is a widely used basal medium for supporting the growth of many different mammalian cells.
$150.00 - $1340.00
Specifications
Content And Storage | Storage conditions: 2°C to 8°C (protect from light) Shipping conditions: Ambient Shelf life: 12 months from date of manufacture |
---|---|
Cell Line | MDCK, glial cells, fibroblasts, human endothelial cells, and rat fibroblasts |
Classification | Animal Origin-free |
Concentration | 1X |
Form | Liquid |
Description
DMEM/F-12 (Dulbecco's Modified Eagle Medium/Nutrient Mixture F-12) is a widely used basal medium for supporting the growth of many different mammalian cells. Cells successfully cultured in DMEM/F-12 include MDCK, glial cells, fibroblasts, human endothelial cells, and rat fibroblasts. We offer a variety of DMEM/F-12 modifications for a range of cell culture applications. Find the right formulation using the media selector tool.
This DMEM/F-12 is modified as follows:
With: L-glutamine, HEPES
Without: Phenol Red
The complete formulation is available.
Using DMEM/F-12
DMEM/F-12 is a 1:1 mixture of DMEM and Ham's F-12. This formulation combines DMEM's high concentrations of glucose, amino acids, and vitamins with F-12's wide variety of components. DMEM/F-12 contains no proteins, lipids, or growth factors. Therefore, DMEM/F-12 may require supplementation, commonly with 10% Fetal Bovine Serum (FBS). DMEM/F-12 uses a sodium bicarbonate buffer system and therefore requires a 5–10% CO2 environment to maintain physiological pH.
Order Info
Shipping Condition: Room Temperature
Specifications
Storage conditions: 2°C to 8°C (protect from light) Shipping conditions: Ambient Shelf life: 12 months from date of manufacture |
|
Animal Origin-free | |
Liquid | |
Standard Serum Supplementation | |
Sterile-filtered | |
No Phenol Red, No Sodium Pyruvate | |
Gibco™ |
MDCK, glial cells, fibroblasts, human endothelial cells, and rat fibroblasts | |
1X | |
DMEM (Dulbecco's Modified Eagle Medium)/F-12 | |
Sterile-filtered | |
High Glucose, Glutamine, HEPES | |
cGMP-compliant under the ISO 13485 standard |
For Research Use Only. Not for use in diagnostic procedures.