missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ DMEM/F-12, no phenol red DFS Item

DMEM/F-12 (Dulbecco's Modified Eagle Medium/Nutrient Mixture F-12) is a widely used basal medium for supporting the growth of many different mammalian cells.

Supplier:  Gibco™ 21041025

Catalog No. 21041025


Only null left
Explore available promotions

Requires : Supplementation

Description

Description

DMEM/F-12 (Dulbecco's Modified Eagle Medium/Nutrient Mixture F-12) is a widely used basal medium for supporting the growth of many different mammalian cells. Cells successfully cultured in DMEM/F-12 include MDCK, glial cells, fibroblasts, human endothelial cells, and rat fibroblasts. We offer a variety of DMEM/F-12 modifications for a range of cell culture applications. Find the right formulation using the media selector tool.

This DMEM/F-12 is modified as follows:

With: L-glutamine

Without: HEPES, Phenol Red

The complete formulation is available.

Using DMEM/F-12

DMEM/F-12 is a 1:1 mixture of DMEM and Ham's F-12. This formulation combines DMEM's high concentrations of glucose, amino acids and vitamins with F-12's wide variety of components. DMEM/F-12 contains no proteins, lipids, or growth factors. Therefore, DMEM/F-12 may require supplementation, commonly with 10% Fetal Bovine Serum (FBS). DMEM/F-12 uses a sodium bicarbonate buffer system and therefore requires a 5–10% CO2 environment to maintain physiological pH.

Order Info

Shipping Condition: Room Temperature

Specifications

Specifications

MDCK, glial cells, fibroblasts, human endothelial cells, and rat fibroblasts
1 X
DMEM (Dulbecco's Modified Eagle Medium)/F-12
Sterile-filtered
No Phenol Red, No HEPES
Gibco
Room Temperature
Animal Origin-free
Liquid
Standard Serum Supplementation
High Glucose, Glutamine, Sodium Pyruvate
cGMP-compliant under the ISO 13485 standard
500 mL
Storage conditions: 2-8° C. Protect from light
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.