Learn More
Gibco™ DPBS (10X), no calcium, no magnesium
Dulbecco's phosphate-buffered saline (DPBS) is a balanced salt solution used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue samples, diluting cells for counting, and preparing reagents.
$114.00 - $984.00
Specifications
Additive | No Sodium Pyruvate |
---|---|
Classification | Animal Origin-Free |
Color | Clear |
Content And Storage | Storage conditions: 15°C to 30°C Shipping conditions: Room temperature Shelf life: 36 months from date of manufacture |
Dilution | 10 X |
Description
Dulbecco's phosphate-buffered saline (DPBS) is a balanced salt solution used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue samples, diluting cells for counting, and preparing reagents. Formulations without calcium and magnesium are required for rinsing chelators from the culture before cell dissociation. Concentrated forms of Gibco™ DPBS require pH adjustment and filtration at the time of preparation (see protocol for details).
We offer a variety of Gibco™ DPBS formulations for a range of cell culture applications.
This DPBS is modified as follows:
Without: Calcium, Magnesium, Phenol Red
The complete formulation is available.
Order Info
Shipping Condition: Room Temperature
Specifications
No Sodium Pyruvate | |
Clear | |
10 X | |
DPBS | |
Mammalian Cell Culture | |
6.7 - 7.0 | |
Room Temperature | |
Liquid |
Animal Origin-Free | |
Storage conditions: 15°C to 30°C Shipping conditions: Room temperature Shelf life: 36 months from date of manufacture |
|
Gibco™ | |
Dulbecco's Phosphate Buffered Saline | |
2630 - 3000 mOsm/kg | |
No Phenol Red | |
Sterile-filtered | |
cGMP-compliant under the ISO 13485 standard |
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.