missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gibco™ DPBS, powder, no calcium, no magnesium DFS Item

Dulbecco's Phosphate-Buffered Saline (DPBS) is a balanced salt solution used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue, diluting cells for counting, and preparing reagents.

Supplier:  Gibco™ 21600044

 View more versions of this product

Catalog No. 21600044


Explore available promotions

Description

Description

Dulbecco's Phosphate-Buffered Saline (DPBS) is a balanced salt solution used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue, diluting cells for counting, and preparing reagents. Formulations without calcium and magnesium are required for rinsing chelators from the culture before cell dissociation. We offer a variety of Gibco™ DPBS formulations for a range of cell culture applications. Find the right formulation using the media selector tool.

This DPBS is modified as follows:

  • Without: Calcium, Magnesium, Phenol Red

The complete formulation is available.

Powder forms of Gibco™ cell culture medium require sodium bicarbonate supplementation, pH adjustment, and filtration at the time of preparation (see protocol for details).

Specifications

Specifications

No Sodium Pyruvate
Storage conditions: 15-30° C
Shipping conditions: Ambient
Shelf life: 36 months from date of manufacture
DPBS
Dulbecco's Phosphate Buffered Saline
265 - 310 mOsm/kg
No Phenol Red
Powder
Animal Origin-Free
Gibco™
50 L
Mammalian Cell Culture
7.2 to 7.7
Room Temperature
cGMP-compliant under the ISO 13485 standard
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.