missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gibco™ DPBS, no calcium, no magnesium DFS Item

Dulbecco's Phosphate-Buffered Saline (DPBS) is a balanced salt solution used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue, diluting cells for counting, and preparing reagents.

Supplier:  Gibco™ 14190235

 View more versions of this product

Catalog No. 14190235


Explore available promotions

Description

Description

Dulbecco's phosphate-buffered saline (DPBS) is a balanced salt solution used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue samples, diluting cells for counting, and preparing reagents. Formulations without calcium and magnesium are required for rinsing chelators from the culture before cell dissociation.

This DPBS is modified as follows:
Without: Calcium, Magnesium, Phenol Red

Specifications

Specifications

No Sodium Pyruvate
Clear
1 X
DPBS
Dulbecco's Phosphate Buffered Saline
Sustainable packaging
7.0 to 7.3
Sterile-filtered
cGMP-compliant under the ISO 13485 standard
Animal Origin-Free
Storage conditions: 15°C to 30°C
Shipping conditions: Room temperature
Shelf life: 36 months from date of manufacture
Gibco™
6 x 1000 mL
Mammalian Cell Culture
270 - 300 mOsm/kg
No Phenol Red
Liquid
Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.