missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gibco™ Amphotericin B DFS Item

Formulation of amphotericin B with sodium deoxycholate as an emulsifying agent to maintain the suspension

$43.95 - $118.00

Specifications

Concentration 0.25 to 2.5 μg/mL
Form Liquid
Product Type Antifungal
For Use With (Application) Prevention of Cell Culture Contamination
Product Line Fungizone™
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
15290018 DFS Item
View Documents
Gibco™
15290018
20 mL
Each for $43.95
 
15290026 DFS Item
View Documents
Gibco™
15290026
50 mL
Each for $118.00
 
Description

Description

 

  • Prevents growth of fungi by causing an increase in fungal plasma membrane permeability
  • Gibco™ Fungizone™ Antimycotic is used to prevent contamination of cell cultures by yeast and multicellular fungi
  • Contains 250μg of amphotericin B and 205μg of sodium deoxycholate per mL of distilled water
  • Recommended working concentration ranges from 0.25 to 2.50μg/mL
Note: For Research use only.    

 

Specifications

Specifications

0.25 to 2.5 μg/mL
Antifungal
Fungizone™
Liquid
Prevention of Cell Culture Contamination
Dry Ice
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

For Research Use Only. Not for use in diagnostic procedures.