missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gibco™ Geneticin™ Selective Antibiotic (G418 Sulfate), Powder DFS Item

Produced by bacterium Micromonospora rhodorangea and acts by binding the ribosome, thus inhibiting protein synthesis in both prokaryotic and eukaryotic cells

Supplier:  Gibco™ 11811031

 View more versions of this product

Catalog No. 11811031


Explore available promotions

Description

Description

Resistance to Geneticin™ Selective Antibiotic (G418 Sulfate) is conferred by the E. coli APH (3) – I and APH (3) – II resistance genes.

  • Used as selective antibiotic in concentration range of 100 - 200μg/mL for bacteria, or 200 – 500μg/mL for most mammalian cells
  • Supplied as powder and should be made into a stock solution of 10 – 50mg/mL in water

Cell Culture, Cloning, Drosophila S2 Cell Culture, High Five™ Cell Culture, Insect Cell Culture, Mammalian Cell Culture, Selection, Sf9 & Sf21 Cell Culture, Transfection, Transformation

Specifications

Specifications

Storage conditions: 15 to 30°C
Shipping conditions: Ambient
Shelf life: 24 months from date of manufacture
10 to 50 μg/mL
Powder
Eukaryotic Selection⁄Stable Cell Line Generation
5 g
Eukaryotic Cells, Prokaryotic Cells
Mammalian Cell Culture, Insect Cell Culture
Antibiotic
Geneticin™
Room Temperature
Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.