missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gibco™ GlutaMAX™ Supplement DFS Item

Gibco GlutaMAX Supplement is an alternative to L-glutamine, with increased stability that improves cell health.

Supplier:  Gibco™ 35050079

 View more versions of this product

Catalog No. 35050079


Explore available promotions

Description

Description

Gibco GlutaMAX Supplement is an alternative to L-glutamine, with increased stability that improves cell health. GlutaMAX Supplement is suitable for both adherent and suspension culture of mammalian cells, with no adaptation required. GlutaMAX Supplement is offered as a 200 mM L-alanyl-L-glutamine dipeptide in 0.85% NaCl. GlutaMAX Supplement is also included in a variety of media formulations. Compared to L-glutamine, GlutaMAX Supplement:

• Minimizes toxic ammonia build-up
• Improves cell viability and growth
• Remains stable across a wide range of temperatures

Minimizes toxic ammonia build-up
Unlike L-glutamine, GlutaMAX Supplement does not spontaneously break down to form ammonia. Instead, cells cleave the dipeptide bond to release L-glutamine as needed. This system prevents waste build-up and maintains a fresh supply of L-glutamine during long-term culture.

Improves cell viability and growth
Cultures with lower levels of ammonia show improved cell viability. In addition, culture conditions are more controlled due to the consistent concentration of GlutaMAX Supplement that is not possible with L-glutamine.

Remains stable across a wide range of temperatures
GlutaMAX Supplement is room-temperature stable and ready to use. GlutaMAX Supplement retains full functionality for 24 months at room temperature, 2 to 8°C, or -5 to -30°C for convenient storage and handling. GlutaMAX Supplement is even stable at 37°C for a week or more.

Specifications

Specifications

Storage conditions: 15°C to 30°C
Alternate storage conditions: -5°C to -20°C and 2°C to 8°C
Shipping conditions: Room temperature
Shelf life: 24 months from date of manufacture
Animal Origin-free
Mammalian Cell Culture
GlutaMax
Clinical Research, Translational Research
cGMP-compliant under the ISO 13485 standard
4.7 to 6
20 x 100 mL
Mammalian Cells
100 X
Liquid
Sterile-filtered
Sustainable packaging
440 - 500 mOsm/kg
Gibco™, GlutaMAX™
Animal origin-free
Videos
SDS
Documents

Documents

Promotions

Promotions