missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gibco™ Ham's F-12 Nutrient Mix DFS Item

Ham's F-12 Nutrient Mixture (F-12) was designed for serum-free, single-cell plating of Chinese Hamster Ovary (CHO) cells.

Supplier:  Gibco™ 11765047

 View more versions of this product

Catalog No. 11765047


Explore available promotions

Requires : Supplementation

Description

Description

Ham's F-12 Nutrient Mixture (F-12) was designed for serum-free, single-cell plating of Chinese Hamster Ovary (CHO) cells. F-12 has since been used for serum-free growth of CHO cultures as well as serum-supplemented growth of other mammalian cells, including chondrocytes and rat prostate epithelial cells. We offer a variety of F-12 modifications for a range of cell culture applications. Find the right formulation using the media selector tool.

This Ham's F-12 is modified as follows:

With: L-glutamine, Phenol Red

The complete formulation is available.

Using F-12

Compared to other basal media, F-12 contains a wider variety of components, including zinc, putrescine, hypoxanthine, and thymidine. F-12 contains no proteins or growth factors. Therefore, F-12 requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). F-12 uses a sodium bicarbonate buffer system (1.176 g/L), and therefore requires a 5–10% CO2 environment to maintain physiological pH.

Order Info

Shipping Condition: Room Temperature

Specifications

Specifications

Storage conditions: 2-8° C. Protect from light
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture
Primary Rat Astrocytes
1 X
Ham's F-12 Nutrient Mixture
Sterile-filtered
No HEPES
Gibco™
Room Temperature
CHO, COS-7, and rat prostate epithelial cells
Animal Origin-free
Liquid
Sterile-filtered
Glutamine, Phenol Red, Sodium Pyruvate
cGMP-compliant under the ISO 13485 standard
1000 mL
Videos
SDS
Documents

Documents

Promotions

Promotions