missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Gibco™ HAT Supplement (50X)
HAT-supplemented medium is suitable for post-fusion selection against unfused or self-fused HGPRT-myeloma cells
Supplier: Gibco™ 21060017
Description
- Liquid mixture of sodium hypoxanthine (5mM), aminopterin (20μM) and thymidine (0.8mM)
- Hypoxanthine and thymidine supply preformed purines and pyrimidines for DNA synthesis by hybridomas via the salvage pathway that utilizes HGPRT contributed by the fused spleen cell
- Aminopterin, a folic acid antagonist, inhibits the de novo nucleoside biosynthesis pathway
Cell Culture, Mammalian Cell Culture
Order Info
Shipping Condition: Dry Ice
Specifications
Storage conditions: -5 to -20°C Shipping conditions: Dry ice Shelf life: 14 months from date of manufacture |
|
50 X | |
HT Supplement | |
6 to 8 | |
Dry Ice |
Mammalian Cells | |
Liquid | |
Sterile-filtered | |
100 mL |
For Research Use Only. Not for use in diagnostic procedures.