missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gibco™ HBSS, calcium, magnesium, no phenol red DFS Item

Hanks' Balanced Salt Solution (HBSS) is used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue samples, diluting cells for counting, and preparing reagents.

Supplier:  Gibco™ 14025126

 View more versions of this product

Catalog No. 14025126


Explore available promotions

Description

Description

Hanks' Balanced Salt Solution (HBSS) is used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue samples, diluting cells for counting, and preparing reagents. Formulations with calcium and magnesium are generally used as transport media, or for reagent preparation.

We offer a variety of Gibco™ HBSS formulations for a range of cell culture applications.

This HBSS is modified as follows:

With: Calcium, Magnesium, Glucose

Without: Phenol Red, Sodium Pyruvate

The complete formulation is available.

Specifications

Specifications

No Sodium Pyruvate
Storage conditions: 15–30°C
Shipping conditions: Ambient
Shelf life: 36 months from date of manufacture
Gibco™
6 x 1000 mL
Mammalian Cell Culture
6.7 - 7.8
Room Temperature
Liquid
Animal Origin-Free
1 X
Hanks' Balanced Salt Solution (HBSS)
Hank's Balanced Salt
270 - 305 mOsm/kg
No Phenol Red
Sterile-filtered
cGMP-compliant under the ISO 13485 standard
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.