missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gibco™ HBSS, no calcium, no magnesium, no phenol red DFS Item

Hanks' Balanced Salt Solution (HBSS) is used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue samples, diluting cells for counting, and preparing reagents.

Supplier:  Gibco™ 14175079

 View more versions of this product

Catalog No. 14175079


Explore available promotions

Description

Description

Hanks' Balanced Salt Solution (HBSS) is used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue samples, diluting cells for counting, and preparing reagents. Formulations without calcium and magnesium are required for rinsing chelators from the culture before cell dissociation.

We offer a variety of Gibco™ HBSS formulations for a range of cell culture applications.

This HBSS is modified as follows:

With: Glucose

Without: Calcium, Magnesium, Phenol Red

The complete formulation is available.

Specifications

Specifications

No Sodium Pyruvate
Storage conditions: 15-30° C
Shipping conditions: Ambient
Shelf life: 36 months from date of manufacture
Gibco™
1000 mL
Mammalian Cell Culture
6.7 - 7.8
Room Temperature
Liquid
Clear
1 X
Hanks' Balanced Salt Solution (HBSS)
Hank's Balanced Salt
270 - 295 mOsm/kg
No Phenol Red
Sterile-filtered
cGMP-compliant under the ISO 13485 standard
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.