missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Gibco™ HEPES (1M)
A zwitterionic organic chemical buffering agent commonly used in cell culture media
$61.75 - $2725.00
Specifications
Chemical Name or Material | Zwitterionic Organic Chemical Buffer |
---|---|
For Use With (Application) | Chromatin Biology |
Physical Form | Liquid |
Solution Type | Buffer |
Sterility | Sterile |
Description
- HEPES (N-2-hydroxyethylpiperazine-N-2-ethane sulfonic acid) is a zwitterionic organic chemical buffering agent commonly used in cell culture media
- The addition of 10–25 mM HEPES provides extra buffering capacity when cell culture requires extended periods of manipulation outside of a CO2 incubator
- A good buffering choice for many cell culture systems because it is membrane impermeable, has limited effect on biochemical reactions, is chemically and enzymatically stable
- It also has very low visible and UV light absorbance
- Store at 2°C to 8°C
Order Info
Shipping Condition: Room Temperature
Specifications
Zwitterionic Organic Chemical Buffer | |
Liquid | |
Sterile |
Chromatin Biology | |
Buffer | |
Biochemical |
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.