missing translation for 'onlineSavingsMsg'
Learn More
S'il vous plaît vous connecter à votre compte en ligne pour afficher votre prix réduits

Gibco™ Horse Serum, heat inactivated, New Zealand origin DFS Item

EIA tested (negative) serum from a donor herd

$39.25 - $134.00

Spécifications

Content And Storage Storage conditions: ≤-10°C, Shipping conditions: Frozen
Species Horse/Equine, Horse
Country of Origin New Zealand
Form Liquid
Product Type Horse Serum
voir les spécifications

Produits 2
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantité  
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantité  
26050070 DFS Item
Afficher les documents
Gibco™
26050070
100 mL
chaque for $39.25
 
26050088 DFS Item
Afficher les documents
Gibco™
26050088
500 mL
chaque for $134.00
 
Description

Description

  • Gibco sera offers excellent value for basic cell culture, specialty research, and specific assays
  • It has earned the trust of researchers with consistent quality and award-winning support
  • Helps meet research needs and budget requirements
  • EIA tested (negative) serum from a donor herd
  • Shipped frozen, store at <10°C
  • Sera Category: Specialty
  • Origin: New Zealand

Order Info

Shipping Condition: Dry ice

Spécifications

Spécifications

Storage conditions: ≤-10°C, Shipping conditions: Frozen
New Zealand
Horse Serum
Gibco™
Horse/Equine, Horse
Liquid
Heat-inactivated
Frozen
Videos
FDS
missing translation for 'documents'

missing translation for 'documents'

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.