missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Gibco™ HT Supplement (100X)
HT-supplemented medium is suitable for post-selection rescue to overcome the effects of residual intracellular aminopterin.
Supplier: Gibco™ 11067030
Description
- Liquid mixture of sodium hypoxanthine (10mM) and thymidine (1.6mM)
- Once the de novo biosynthesis pathway for nucleosides has been reestablished, HT supplementation can be discontinued
- Manufactured at a cGMP compliant facility, located in Grand Island, New York, registered with the FDA as a medical device manufacturer and certified to ISO 13485 standards
Order Info
Shipping Condition: Room Temperature
Specifications
Storage conditions: 2°C to 8°C Shipping conditions: Room temperature Shelf life: 18 months from date of manufacture |
|
Liquid | |
Sterile-filtered |
100 X | |
HT Supplement | |
Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.