missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Gibco™ Hybridoma-SFM, powder
Complete, ready-to-use formulation
Supplier: Gibco™ 12300067
Description
Gibco Hybridoma SFM is a serum-free, very low-protein medium specifically developed for the ability to support the growth of hybridomas for antibody production.
- Ability to support suspension and stationary hybridomas and myelomas
- Serum-free, very low-protein formulation
- Supports suspension and stationary hybridomas and myelomas
- Suitable for the culture of recombinant myeloma lines as well as traditional hybridomas
- Provides high cell densities and IgG production
- Cholesterol-dependent cultures such as NS0 require the addition of 250X Cholesterol Lipid Concentrate
- Complete, ready-to-use formulation
- No supplementation is required
- Serum-free, very low-protein formulation, allowing for easier purification of your protein of interest
- Contains < 20μg/mL total protein
Antibody and Cell Culture Production, Bioproduction, Cell Culture, Mammalian Cell Culture
Order Info
Shipping Condition: Room temperature
Specifications
Storage conditions: 2-8°C Shipping conditions: Ambient Shelf life: 33 months from date of manufacture |
|
Low Protein, Serum-free | |
Hybridoma Medium, powder | |
Glutamine | |
10 L |
Hybridomas | |
Powder | |
Serum-free | |
Gibco™ | |
Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.