missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gibco™ IMDM DFS Item

IMDM (Iscove's Modified Dulbecco's Medium) is well suited for rapidly proliferating, high-density cell cultures, including Jurkat, COS-7, and macrophage cells

Supplier:  Gibco™ 12440046

 View more versions of this product

Catalog No. 12440046


Explore available promotions

Requires : Supplementation

Description

Description

IMDM (Iscove's Modified Dulbecco's Medium) is well suited for rapidly proliferating, high-density cell cultures, including Jurkat, COS-7, and macrophage cells. We offer a variety of IMDM modifications for a range of cell culture applications. Find the right formulation using the media selector tool.

This IMDM is modified as follows:

With: L-glutamine, Phenol Red

Without: α-thioglycerol, 2-mercaptoethanol

The complete formulation is available.

Using IMDM

IMDM, a modification of Dulbecco's Modified Eagle Medium, includes selenium as well as additional amino acids and vitamins. In addition, this unique medium lacks iron, with potassium nitrate replacing ferric nitrate. IMDM contains no proteins, lipids, or growth factors. Therefore, IMDM requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). IMDM uses a sodium bicarbonate buffer system (3.024 g/L), and therefore requires a 5–10% CO2 environment to maintain physiological pH.

Order Info

Shipping Condition: Room Temperature

Specifications

Specifications

Jurkat, COS-7, and macrophage cells
1 X
IMDM (Iscove's Modified Dulbecco's Medium)
Sterile-filtered
No α-Thioglycerol, No 2-Mercaptoethanol
Gibco™
Room Temperature
Animal Origin-free
Liquid
Sterile-filtered
High Glucose, Glutamine, HEPES, Phenol Red, Sodium Pyruvate
cGMP-compliant under the ISO 13485 standard
1000 mL
Storage conditions: 2-8° C. Protect from light
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

For in vitro diagnostic use.