Gibco™ Kanamycin Sulfate DFS Item
Water-soluble antibiotic originally purified from bacterium Streptomyces kanamyceticus
Supplier:  Gibco™ 11815032
 View more versions of this product
Catalog No. 11815032
Explore available promotions
Description
Description
  • A water-soluble antibiotic originally purified from the bacterium Streptomyces kanamyceticus
  • Kanamycin acts by binding to the 30S subunit of the bacterial ribosome and inhibiting protein synthesis in susceptible bacteria
  • Gibco™ Kanamycin is effective against a wide variety of gram-negative and some gram-positive bacteria
  • It is used for the prevention of bacterial contamination of cell cultures
  • Provided as a 10 mg/mL stock solution, the recommended working concentration for cell culture is 100 μg/mL
  • Manufactured at a cGMP compliant facility that is registered with the FDA as a medical device manufacturer and certified to ISO 13485 standards
  • Store at -5°C to -20°C
For Research Use Only: Not intended for animal or human diagnostic or therapeutic use.
Specifications
Specifications
Store at room temperature.
Antibiotic
25 g
Powder
Bacterial Selection
Ambient Temperature
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
For Research Use Only. Not for use in diagnostic procedures.