missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gibco™ Kanamycin Sulfate DFS Item

Water-soluble antibiotic originally purified from bacterium Streptomyces kanamyceticus

Supplier:  Gibco™ 15160054

 View more versions of this product

Catalog No. 15160054


Explore available promotions

Description

Description

  • A water-soluble antibiotic originally purified from the bacterium Streptomyces kanamyceticus
  • Kanamycin acts by binding to the 30S subunit of the bacterial ribosome and inhibiting protein synthesis in susceptible bacteria
  • Gibco™ Kanamycin is effective against a wide variety of gram-negative and some gram-positive bacteria
  • It is used for the prevention of bacterial contamination of cell cultures
  • Provided as a 10 mg/mL stock solution, the recommended working concentration for cell culture is 100 μg/mL
  • Manufactured at a cGMP compliant facility that is registered with the FDA as a medical device manufacturer and certified to ISO 13485 standards
  • Store at -5°C to -20°C

For Research Use Only: Not intended for animal or human diagnostic or therapeutic use.

Specifications

Specifications

Storage conditions: -5 to -20°C
Shipping conditions: Frozen
Shelf life: 12 months from date of manufacture
Liquid
Sterile-filtered
Bacterial Selection
Dry Ice
100 X, 10 mg/mL
Antibiotic
Sterile-filtered
100 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.