Learn More
Gibco™ L-Glutamine (200 mM)
Ready-to-use stock solution that acts as a cell culture supplement
$26.56 - $1214.00
Specifications
Chemical Name or Material | Glutamine |
---|---|
pH | 5 to 6 |
Concentration or Composition (by Analyte or Components) | 100X |
Physical Form | Liquid |
Description
L-Glutamine is an amino acid that is required for cell culture. L-Glutamine participates in the formation of purine and pyrimidine nucleotides, amino sugars, glutathione, L-glutamate, and other amino acids, as well as in protein synthesis and glucose production. Unlike most other amino acids, L-glutamine is not stable in solution. The rate at which degradation proceeds is a function of time, temperature, and pH.
- Optimal concentration is dependent upon the cell type and medium used to culture the cells, but generally falls in the range of 2 to 6 mM
- Manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK, registered with the FDA as a medical device manufacturer and certified to the ISO 13485 standard
Bioproduction, Cell Culture, Cloning, Mammalian Cell Culture, Transformation, Viral Vaccine Production
Order Info
Shipping Condition: Dry Ice
Specifications
Glutamine | |
5 to 6 | |
100X | |
Liquid |