missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Leibovitz's L-15 Medium DFS Item

Leibovitz's L-15 Medium supports the growth of HEP-2 monkey kidney cells and primary explants of embryonic and adult human tissue.

$107.00 - $956.00

Specifications

Cell Line HEP-2 monkey kidney cells
Classification Animal Origin
Concentration 1 X
Form Liquid
Product Type Leibovitz's L-15 Medium
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
11415064 DFS Item
View Documents
Gibco™
11415064
500 mL
Each for $107.00
Only null left
 
11415114 DFS Item
View Documents
Gibco™
11415114
10 x 500 mL
Each for $956.00
Only null left
 
Description

Description

Leibovitz's L-15 Medium supports the growth of HEP-2 monkey kidney cells and primary explants of embryonic and adult human tissue. L-15 is buffered by phosphates and free base amino acids instead of sodium bicarbonate. This medium is designed for supporting cell growth in environments without CO2 equilibration.

This L-15 is modified as follows:

With: Galactose, Phenol Red, L-glutamine, Sodium Pyruvate

Without: Glucose, HEPES, Sodium Bicarbonate

The complete formulation is available.

L-15 contains no proteins, lipids, or growth factors. Therefore, F-10 requires supplementation, commonly with 10% Fetal Bovine Serum (FBS).

Order Info

Shipping Condition: Room Temperature

Specifications

Specifications

HEP-2 monkey kidney cells
1 X
Leibovitz's L-15 Medium
Sterile-filtered
No Glucose, No HEPES, No Sodium Bicarbonate
Gibco
Storage conditions: 2-8° C. Protect from light
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture
Animal Origin
Liquid
Sterile-filtered
Glutamine, Phenol Red, Sodium Pyruvate, Galactose
cGMP-compliant under the ISO 13485 standard
Room Temperature
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.