Learn More
Gibco™ MEM α, GlutaMAX™ Supplement, no nucleosides
Minimum Essential Medium (MEM) α is widely used for mammalian cell culture as well as selection for transfected DHFR negative cells. MEM α can be used with a variety of suspension and adherent mammalian cells, including keratinocytes, primary rat astrocytes, and human melanoma cells.
$117.00 - $984.00
Specifications
| Classification | Animal Origin-free |
|---|---|
| Concentration | 1 X |
| Form | Liquid |
| Product Type | MEM α (Minimum Essential Medium α) |
| Serum Level | Standard Serum Supplementation |
Description
Minimum Essential Medium (MEM) α is widely used for mammalian cell culture as well as selection for transfected DHFR negative cells. MEM α can be used with a variety of suspension and adherent mammalian cells, including keratinocytes, primary rat astrocytes, and human melanoma cells. We offer a variety of Gibco™ MEM α modifications for a range of cell culture applications. Find the right formulation using the media selector tool.
This MEM α is modified as follows:
With: Phenol Red, GlutaMAX™
Without: Ribonucleosides, Deoxyribonucleosides
The complete formulation is available.
Gibco™ MEM α is a modification of Minimum Essential Medium (MEM) that contains non-essential amino acids, sodium pyruvate, lipoic acid, vitamin B12, biotin, and ascorbic acid. MEM α is available without nucleosides for use as a selection medium for DG44 and other DHFR negative cells. Gibco™ MEM α with GlutaMAX™ supplement minimizes toxic ammonia build-up and improves cell viability and growth in an easy-to-use format. This product is made with Earle's salts.
MEM α contains no proteins, lipids, or growth factors. Therefore, MEM α requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). MEM α uses a sodium bicarbonate buffer system (2.2 g/L) and therefore requires a 5-10% CO2 environment to maintain physiological pH.
Order Info
Shipping Condition: Room Temperature
Specifications
| Animal Origin-free | |
| Liquid | |
| Standard Serum Supplementation | |
| Sterile-filtered | |
| cGMP-compliant under the ISO 13485 standard |
| 1 X | |
| MEM α (Minimum Essential Medium α) | |
| Sterile-filtered | |
| No HEPES, No Ribonucleosides, No Deoxyribonucleosides | |
| Gibco, GlutaMAX |
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.