Learn More
Gibco™ MEM α, nucleosides, no phenol red
Minimum Essential Medium (MEM) α is widely used for mammalian cell culture as well as selection for transfected DHFR negative cells.
$95.50 - $860.00
Specifications
Content And Storage | Storage conditions: 2-8° C. Protect from light Shipping conditions: Ambient Shelf life: 12 months from date of manufacture |
---|---|
Cell Line | HeLa, BHK-21, 293, HEP-2, HT-1080, MCF-7, and fibroblasts |
Cell Type | Primary Rat Astrocytes |
Classification | Animal Origin-free |
Concentration | 1 X |
Description
Minimum Essential Medium (MEM) α is widely used for mammalian cell culture as well as selection for transfected DHFR negative cells. MEM α can be used with a variety of suspension and adherent mammalian cells, including keratinocytes, primary rat astrocytes, and human melanoma cells. We offer a variety of Gibco™ MEM α modifications for a range of cell culture applications. Find the right formulation using the media selector tool.
This MEM α is modified as follows:
With: Ribonucleosides, Deoxyribonucleosides, L-glutamine
Without: Phenol Red
The complete formulation is available.
Gibco™ MEM α is a modification of Minimum Essential Medium (MEM) that contains non-essential amino acids, sodium pyruvate, lipoic acid, vitamin B12, biotin, and ascorbic acid. MEM α is available without nucleosides for use as a selection medium for DG44 and other DHFR negative cells. This product is made with Earle's salts.
Product Use
For Research Use Only: Not intended for animal or human diagnostic or therapeutic use.
MEM α contains no proteins, lipids, or growth factors. Therefore, MEM α requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). MEM α uses a sodium bicarbonate buffer system (2.2 g/L) and therefore requires a 5-10% CO2 environment to maintain physiological pH.
Order Info
Shipping Condition: Room Temperature
Compliance
Gibco MEM α is manufactured at a cGMP compliant facility located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.
Specifications
Storage conditions: 2-8° C. Protect from light Shipping conditions: Ambient Shelf life: 12 months from date of manufacture |
|
Primary Rat Astrocytes | |
1 X | |
MEM α (Minimum Essential Medium α) | |
Sterile-filtered | |
No HEPES, No Phenol Red | |
Gibco™ |
HeLa, BHK-21, 293, HEP-2, HT-1080, MCF-7, and fibroblasts | |
Animal Origin-free | |
Liquid | |
Standard Serum Supplementation | |
Low Glucose, Glutamine, Sodium Pyruvate, Ribonucleosides, Deoxyribonucleosides | |
cGMP-compliant under the ISO 13485 standard | |
Room Temperature |
For Research Use Only: Not intended for animal or human diagnostic or therapeutic use.