missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gibco™ Newborn Calf Serum, heat inactivated, New Zealand origin DFS Item

Secure sera for cell culture with least viral risk sera from BSE-free regions

$64.00 - $239.00

Specifications

Content And Storage Storage conditions: ≤-10°C
Shipping conditions: Frozen
Age Newborn (14 days or less)
Species Cattle/Bovine
Country of Origin New Zealand
Form Liquid
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
26010066 DFS Item
View Documents
Gibco™
26010066
100 mL
Each for $64.00
 
26010074 DFS Item
View Documents
Gibco™
26010074
500 mL
Each for $239.00
 
Description

Description

  • Sera Category: Secure
  • Origin: New Zealand
  • Collected from calves 14 days old or less

Cell Culture, Mammalian Cell Culture

Order Info

Shipping Condition: Dry Ice

Specifications

Specifications

Storage conditions: ≤-10°C
Shipping conditions: Frozen
Cattle/Bovine
Liquid
Newborn Calf Bovine Serum
Gibco™
Newborn (14 days or less)
New Zealand
Bottle
Heat-inactivated
Frozen
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

For In Vitro Diagnostic Use.