Learn More
Gibco™ PBS (10X), pH 7.4 DFS Item
Phosphate-buffered saline (PBS) is a balanced salt solution that is used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue samples, diluting cells for counting, and preparing reagents.
Supplier:  Gibco™ 70011044
 View more versions of this product
Catalog No. 70011044
Only null left
Explore available promotions
Description
Description
Phosphate-buffered saline (PBS) is a balanced salt solution that is used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue samples, diluting cells for counting, and preparing reagents. PBS is formulated without calcium and magnesium for rinsing chelators from the culture before cell dissociation. Concentrated forms of Gibco™ PBS may require pH adjustment at the time of dilution (see protocol for details).
We offer a variety of Gibco™ PBS formulations for a range of cell culture applications.
This PBS is manufactured as follows:
Without: Calcium, Magnesium, Phenol Red
The complete formulation is available.
Specifications
Specifications
Phosphate Buffered Saline (PBS)
10 X
7.4
Gibco
Phosphate Buffered Saline
No Sodium Pyruvate
2800 - 3100 mOsm/kg
White
Mammalian Cell Culture
Liquid
500 mL
Sterile-filtered
cGMP-compliant under the ISO 13485 standard
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.